β-Glycerophosphate disodium salt hydrate ≥99% (by titrimetric analysis)

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: Disodium-β-glycerophosphate hydrate, Glycerol 2-phosphate disodium salt hydrate, Sodium 1,3-dihydroxypropan-2-yl phosphate hydrate(2:1:x), BGP

01454-100G 01454-25G 01454-5G
CH01454-100GEA 153.89 CAD
CH01454-100G CH01454-25G CH01454-5G
β-Glycerophosphate disodium salt hydrate ≥99% (by titrimetric analysis)
β-Glycerophosphate disodium salt hydrate

Used as an organic phosphate donor and to buffer cell culture media.

Formula: C₃H₇Na₂O₆P·H₂O
MW: 234.05
Melting Pt: 102...104 °C
Storage Temperature: Ambient
MDL Number: MFCD00149082
CAS Number: 154804-51-0

Order Now

Specification Test Results

Appearance White Crystalline Powder
Heavy Metals ≤20 ppm
Loss on Drying 26.0 - 32.0%
Assay by Titration (Potentiometric) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR