Tetrakis(4-aminophenyl)methane ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4',4'',4'''-Methanetetrayltetraaniline

T2947-1G T2947-200MG
TCT2947-1GEA 421.73 CAD
TCT2947-1G TCT2947-200MG
Tetrakis(4-aminophenyl)methane ≥95.0% (by HPLC, titration analysis)
Tetrakis(4-aminophenyl)methane
Formula: C₂₅H₂₄N₄
MW: 380.5 g/mol
Melting Pt: 320 °C (dec., lit.)
MDL Number: MFCD12546930
CAS Number: 60532-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f279f664bdbb86d9194b543b74e81bce69b1a4a&vid=1f279f664bdbb86d9194b543b74e81bce69b1a4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) Min. 95.0%
Purity(Nonaqueous Titration) Min. 95.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR