1-Naphtholbenzeine

Synonyms: alpha-Naphtholbenzein, p-Naphtholbenzein, α-Naphtholbenzein, p-Naphtholbenzeine, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one, 1-Naphtholbenzein, Bis(4-hydroxy-1-naphthyl)phenylmethanol, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol, α-Naphtholbenzeine

22928-25G 22928-5G
CH22928-25GEA 139.4 CAD
CH22928-25G CH22928-5G
1-Naphtholbenzeine
1-Naphtholbenzeine
Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Melting Pt: 230...235 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6

Order Now

Specification Test Results

Appearance Brown Powder
Visual Transition Range From pH 8.0 (Reddish Orange) to pH 10.0 (Greenish Blue)
Loss on Drying ≤7.0% (105 °C)
Specific Absorption 1% on Dried Basis at 626 nm ≥640
Absorption Maximum 622 - 626 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR