Chemware® PFA Test Tubes, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069598 D1069599 D1069695
66111-574EA 227.01 CAD
66111-574 66111-578 66111-564
Chemware® PFA Test Tubes, Saint-Gobain Performance Plastics
Tubes Test Tubes
These unbreakable test tubes are excellent for trace metal analysis.
  • Made from fluoropolymer PFA resin
  • Low metal ion content
  • Inert
They are non-contaminating, and transparent for easy viewing of contents. They withstand extreme temperature. Can be sterilized repeatedly.