Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
EL5 EL10 EL20 EL30 EL60 EL1 EL3
76290-380BX 43.04 CAD
76290-380 76290-382 76290-384 76290-386 76290-388 76290-420 76290-422
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Syringes General-Purpose Syringes
EXEL international's quality disposable syringes. Great value at reasonable costs.
  • Standard three part syringe
  • Latex and pyrogen free
  • Non-toxic
  • Positive plunger stop
EXEL international is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.
Certifications: Meets ISO standards, CE registered.
Packaging: Sterile, individually wrapped.