Interchangeable Glass Syringe with Luer Lock Tip, Ace Glass Incorporated
Supplier: Ace Glass
13681-02 13681-05 13681-10 13681-20 13681-30 13681-50
76312-822EA 242.07 CAD
76312-822 76312-824 76312-826 76312-828 76312-830 76312-832
Interchangeable Glass Syringe with Luer Lock Tip, Ace Glass Incorporated
Syringes General-Purpose Syringes
Interchangeable syringe featuring a Luer lock tip, permanently fused graduations, and a colored plunger end for easy and accurate dosage measurement.
  • Made of high-quality borosilicate glass
  • Reinforced flanges that are flat on two sides to prevent rolling
  • Marked for non-medical use; no medical license required
Manufactured to ensure the perfect mating of barrels and plungers, so any interchangeable plunger of a particular size will fit any interchangeable barrel of the same size.
Order Now
SPECIFICATIONS
Environmentally Preferable