Quick-DNA™ Kits, Zymo Research
Supplier: Zymo Research
D3010 D3011 D3012 D3020 D3021 D3024 D3025
77001-076EA 422.6 CAD
77001-076 77001-078 77001-080 77001-090 77001-092 77001-094 77001-096
Quick-DNA™ Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
DNA from cells, swabs, and whole blood.
  • Quick & easy: Simple 20 minute procedure
  • Unique technology excludes the use of proteinase K and organic denaturants
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more
The Quick-DNA™ kits are ideal for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin™ column/plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.