Pre-Printed Laboratory Warning Labels
Supplier: GA INTERNATIONAL INC
807656026503 807656026527 807656026497 807656083940 807656083971 807656026565 807656026589 807656026596 807656026602 807656026619 807656026626 807656084077 807656084084
76275-958RL 59.83 CAD
76275-958 76275-962 76275-964 76276-104 76276-108 76276-112 76276-114 76276-116 76276-120 76276-122 76276-124 76276-126 76276-128 76276-130 76276-132 76276-134 76276-136
Pre-Printed Laboratory Warning Labels
Labels
Various pre-printed laboratory warning labels designed to advise staff and visitors of potential hazards and dangers. This includes biohazard and radioactive warning labels, as well as standardized THC warning labels for labeling a wide range of cannabis products.
  • Temperature range: −80 to +100 °C (−112 to +212 °F)
  • Withstands ultra-low temperature conditions (−80 °C/−112 °F)
  • Special construction protects against abrasion, chemical exposure, and tearing
  • Durable and high-quality materials for long-lasting use
Pre-printed general warning labels forMore Product Information