Standard Opening Crimp Top Vials, Chemglass
Supplier: Chemglass
CV-2625-1232 CV-2615-1232 CV-2620-1232 CV-1170-1232 CV-1171-1232 CV-1168-1232 CV-1169-1232
10171-042PK 162.07 CAD
10171-042 10171-044 10171-046 75819-712 75819-714 75820-626 75820-628
Standard Opening Crimp Top Vials, Chemglass
Vials
These vials are designed to work with autosamplers incorporating a robotic arm as well as other autosamplers.
  • Suitable for poly Crimp™ seals or standard aluminum seals
  • Available with or without graduated marking spots
Vials incorporates the unique step vial design that precisely centers a limited volume insert in the vial neck.
Order Now
SPECIFICATIONS
Environmentally Preferable