Extended Exposure Xylene and Chemical-Resistant Thermal Transfer Labels
Supplier: GA INTERNATIONAL INC
807656028958 807656028965 807656019178 807656019185 807656019192 807657642627 807657642634 807656084725 807656084756 807656084824 807656084923 807656088389 807656098869 807656099026 807656101200 807656102726 807656103693
76210-324RL 152.52 CAD
76210-324 76210-326 76210-332 76210-608 76274-404 76274-406 76274-408 76274-410 76274-412 76274-414 76274-416 76274-418 77321-324 77321-326 77321-304 77321-306 77321-308 77321-310 77321-312 77321-314 77321-316 77321-318 77321-320 77321-322
Extended Exposure Xylene and Chemical-Resistant Thermal Transfer Labels
Labels
Xylene and chemical-resistant labels for the identification of microscope slides that will undergo long-term chemical exposure and immersion.
  • Temperature range: –80 to +120 ℃ (–112 to +248 ℉)
  • Excellent stain resistance, ideal for microscope slides used in histology
  • Recommended for routine as well as special staining protocols
  • Withstand exposure to xylene, toluene, hexane, acetone, alcohols, dimethyl sulfoxide (DMSO), methyl ethyl ketone (MEK), acetonitrile (ACN), and other common solvents
  • Resistant to the new generation of safer, non-toxic, xylene
More Product Information