Serological Pipettes, Class A
Supplier: Wards
CH0573C CH0573D CH0573E CH0573A CH0573B
470312-900EA 16.45 CAD
470312-900 470312-902 470312-904 470312-924 470312-926
Serological Pipettes, Class A
Pipettes Serological Pipettes
Perfect pipette for everyday laboratory use.
  • Reusable
  • Color-coded for easy sorting
  • Accuracy as per Class A and DIN ISO 835 standard
These class A serological pipettes are calibrated for delivery to jet. The last drop is expelled by blowing. Blue graduations and color coding make these pipettes very easy to use.
Order Now
SPECIFICATIONS
Environmentally Preferable