LpH® III st Disinfectant Cleaners
Supplier: Steris
LpH® III
1S1829 1S1808 1S16CX 1S1829CA
76303-142CS 1547.75 CAD
76303-142 CA76303-142 76303-144 76303-140
LpH® III st Disinfectant Cleaners
Cleaning Agents Disinfectants
STERIS' LpH® III st Phenolic disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Gamma irradiated and double bagged for easy introduction into your aseptic areas
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' LpH® III st Disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the LpH® III disinfectants kill a broad spectrum of common bacteria and fungi. The One Solution st LpH® III System Pre-measured WFI combines a sterile unit dose pouch with a gallon of USP Water For Injection (WFI). The acidic LpH® III Disinfectant formulation can be used alone or in rotation with the Vesphene® III Disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS Technical services team for individualized application and validation assistance.