Urine Collection System, Simport Scientific
Supplier: Simport Scientific
T410-1 T410 T410-2 T410-3 T410-2 T410-3
CA15705-351CS 205.14 CAD
CA15705-351 CA53594-089 CA73960-322 CA73960-320L 76299-290 76299-292
Urine Collection System, Simport Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
The urine collection system is designed to allow the use of midget urinometers and reagent test strips requiring only 0.25 or 0.5 ml of sample.
  • Tube is made of virgin polystyrene
  • Flared tube tops allow for easy filling
  • Drip-free pouring
  • Tube dimensions: 105×21 mm (Hר)
Urine tubes are made of virgin polystyrene and are free of any mold release agents, metals, or additives that could contaminate samples. The heavy-wall tubes are graduated at 0.25, 0.5, 1, 2 and every 2 ml thereafter up to 12 ml. They can be safely centrifuged at speeds up to 2000×g. The tubes are flared at the top to make filling and drip-free pouring easier. The tight-fitting plastic caps makes these tubes suitable for transportation in pneumatic tube systems.
Delivery information: The urine collection system contains 100 disposable 15 ml heavy-wall polystyrene tubes, snap caps, self-adhesive identification labels, and 3 oz. plastic collection cups all packed in a plastic bag (5 bags per case).