Cryogenic Labels for Identification of IVF Straws
Supplier: GA INTERNATIONAL INC
807656031644 807656031651 807656031668 807656031675 807656019697 807656019703 807656019710 807656019727 807656019734 807656019741 807656019758 807656019765 807656019772 807656019789 807656019796 807656019802 807656019819 807656019826 807656019833 807656019840 807656019857 807656019864 807656019871 807656019888 807656019895 807656019901
76210-238RL 107.74 CAD
76210-238 76210-474 76210-476 76210-478 76274-516 76274-518 76274-520 76274-522 76274-524 76274-526 76274-528 76274-530 76274-532 76274-534 76274-536 76274-538 76274-540 76274-542 76274-544 76274-546 76274-548 76274-550 76274-552 76274-554 76274-556 76274-558
Cryogenic Labels for Identification of IVF Straws
Labels
These labels are designed for identifying vitrification straws and devices used in in-vitro fertilization (IVF) clinics and ART facilities. These cryogenic labels are compatible with various sizes of high-security straws and vitrification devices.
  • Temperature range: −196 to +80 °C (−321 to 176 °F)
  • Ideal for short and long-term storage in cryogenic conditions, including liquid nitrogen (−196 °C)
  • Specifically designed for the identification of cryo-preservation straws (0.25 and 0.5 ml) used in IVF laboratories
  • Printable with all major thermal transfer
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable