Xylene and Chemical-Resistant Labels (Stain-Proof/Resin-Embedding)
Supplier: GA INTERNATIONAL INC
807656044088 807656044095 807656031576 807656031569 807657613719 807656019291 807656019307 807656019314 807656019321 807656019260 807656019277 807656005256 807656019284 807656019338 807656019345 807656019352 807656019369 807656019376 807656006185 807656019383 807656029191 807656028019 807656019390 807656019406 807656019413 807656019420 807656028514 807656029207 807656029214 807656029177 807656029184 807656019437 807656019444 807656019451 807656019468
76210-086RL 156.63 CAD
76210-086 76210-088 76210-382 76210-470 76274-284 76274-286 76274-288 76274-290 76274-292 76274-342 76274-344 76274-346 76274-348 76274-422 76274-424 76274-426 76274-428 76274-430 76274-432 76274-434 76274-436 76274-438 76274-440 76274-442 76274-444 76274-446 76274-448 76274-452 76274-454 76274-456 76274-458 76274-460 76274-462 76274-464 76274-466
Xylene and Chemical-Resistant Labels (Stain-Proof/Resin-Embedding)
Labels
Designed for histological staining protocols and resin embedding. These thermal transfer printable labels offer excellent chemical resistance and withstand a wide range of temperatures.
  • Temperature range: –80 to +149 °C (–112 to +300 °F)
  • Able to withstand exposure to harsh chemicals and solvents
  • Printable with all major thermal transfer printer brands
  • Perfect for labeling microscope slides used throughout the histological staining process
  • Resistant to gamma irradiation (tested up to 50 KGy)
  • Chemical-resistant labels withstand embedding in histological
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable