Exel International® Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
EN181 EN18112 EN191 EN19112 EN201 EN27112 EN2834 EN3012 EN301 EN20112 EN211 EN21112 EN2234 EN221 EN22112 EN2334 EN231 EN23112 EN2558 EN251 EN25112 EN2612 EN2712 EN27114
76290-474BX 14.34 CAD
76290-474 76290-476 76290-478 76290-480 76290-482 76290-484 76290-486 76290-488 76290-490 76290-548 76290-550 76290-552 76290-554 76290-556 76290-558 76290-560 76290-562 76290-564 76290-566 76290-568 76290-570 76290-572 76290-574 76290-576
Exel International® Hypodermic Needles, Air-Tite Products
Syringe Needles
High quality, inexpensive hypodermic needles from EXEL international.
  • Individually sterilized
  • Pyrogen free
  • 304 stainless steel
  • Needles and boxes are color coded
EXEL international offers a wide range of hypodermic needles for human, laboratory and veterinary use. These needles are siliconized for smoother gliding during injection, and have a durable regular thickness wall.
Certifications: ISO certified, CE certified
Packaging: Sterile, individually wrapped.