Ethanol grain derived 200 Proof

Supplier: Ricca Chemical
Danger

Synonyms: Ethyl alcohol

RSOE0013-55E RSOE0013-5E4
RCRSOE0013-55EEA 6754.42 CAD
RCRSOE0013-55E RCRSOE0013-5E4
Ethanol grain derived 200 Proof
Ethanol
Formula: C₂H₅OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Acidity (as Acetic Acid) max 0.003
Alkalinity (as NH3) max 3
Appearance clear, colorless
Assay min 94.9 % (v/v)
Fusel Oil To Pass Test
Ketones/Isopropanol To Pass Test
Lead (Pb) max 0.50 mg/Kg
Methanol (pass/fail) To Pass Test
Nonvolatile Residue max 0.003 %
Odor To Pass Test
Solubility in Water To Pass Test
Specific Gravity @ 15.56°C max 0.8161
Specific Gravity @ 25.0°C max 0.8096
Substances Darkened by Sulfuric Acid To Pass Test
Substances Reducing Permanganate To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR