YMC Hydrosphere C18 HPLC Columns, YMC America
Supplier: YMC America,Inc
HS12S02-0302WT HS12S02-0303WT HS12S02-0502WT HS12S02-0503WT HS12S02-0546WT HS12S02-1002WT HS12S02-1003WT HS12S02-L502WT HS12S02-L503WT HS12S03-0501WT HS12S03-0502WT HS12S03-0503WT HS12S03-0506WT HS12S03-0546WT HS12S03-1001WT HS12S03-1002WT HS12S03-1003WT HS12S03-1006WT HS12S03-1046WT HS12S03-1502WT HS12S03-1503WT HS12S03-1546WT HS12S03-2546WT HS12S03-H501WT HS12S03-H502WT HS12S03-H546WT HS12S03-L502WT HS12S03-L506WT HS12S03-L546WT HS12S05-0502WT HS12S05-0503WT HS12S05-0546WT HS12S05-1002WT HS12S05-1003WT HS12S05-1046WT HS12S05-1501WT HS12S05-1502WT HS12S05-1503WT HS12S05-1506WT HS12S05-2501WT HS12S05-2502WT HS12S05-2503WT HS12S05-2506WT HS12S05-2546WT HS12S05-H502WT HS12S05-L502WT HS12S05-L503WT HS12S05-L546WT
97030-912EA 2288.72 CAD
97030-912 97030-914 97030-916 97030-918 97030-920 97030-922 97030-924 97030-926 97030-928 97030-940 97030-942 97030-944 97030-946 97030-948 97030-950 97030-952 97030-954 97030-956 97030-958 97030-960 97030-962 97030-964 97030-966 97030-968 97030-970 97030-976 97030-978 97030-980 97030-982 97030-984 97030-986 97030-988 97031-002 97031-004 97031-010 97031-012 97031-014 97031-020 97031-022 97031-024 97031-026 97031-028 97031-036 97031-038 97031-040 97031-042 97031-044 97031-050 97031-052 97031-058 97031-060 97031-062 97031-064
YMC Hydrosphere C18 HPLC Columns, YMC America
Chromatography Columns
These hydrosphere C18 columns are made with the combination of ultra-pure silica and unique selectivity which leads to a stationary phase that fulfills the relevant requirements of HPLC users.
  • Based on highly inert, ultra-pure, pH neutral silica
  • Hydrophilic C18 surface for enhanced polar selectivity
  • Extremely rigorous physical and chemical specifications
  • Suitable for conventional reversed phase and highly aqueous applications
Minimal metal content and very low concentrations of residual silanols make this an ideal phase for the analysis of basic pharmaceuticals and chelating compounds even under LC-MS conditions.
In added to that the possibility to separate polar compounds under highly aqueous mobile phase conditions without loss in retention, plate counts, and peak performance greatly enhances the versatility of these columns.