KIMBLE® HI-VAC® Right-Angle Valve, with Glass Plug and Tip O-Rings, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
826460-0004 826460-0008 826460-0012
KT82640-0004CS 221.2 CAD
KT82640-0004 KT826460-0008 KT826460-0012
KIMBLE® HI-VAC® Right-Angle Valve, with Glass Plug and Tip O-Rings, DWK Life Sciences
Valves Vacuum Valves
This right-angle valve uses a glass plug (as an alternative to PTFE or CTFE) for applications where the 'out-gassing' plug material is undesirable.
  • All plugs are totally interchangeable with other HI-VAC® counterparts
  • Greater solvent resistance
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with FKM O-rings.