COUNT-CAL™ Count Precision Size Standards, Microgenics

Supplier: MICROGENICS CORP (FKA DUKE) MS

CC02-PK CC05-PK CC10-PK
MSPP-CC02PKEA 513.85 CAD
MSPP-CC02PK MSPP-CC05PK MSPP-CC10PK
COUNT-CAL™ Count Precision Size Standards, Microgenics
Standards Colour Reference and Spectroscopy Standards

COUNT-CAL™ count precision size standards, are easy to use product for validating liquid particle counters.


  • Cost-effective, convenient
  • World class particle technology solutions
  • Polystyrene microspheres are suspended in an ultra clean diluent
  • Concentration is optimized for use in light obscuration particle counters and other low concentration counting applications


Minimize contamination by eliminating the need for serial dilutions and extensive sample handling


These standards are designed for regular use in liquid particle counters to verify instrument calibration before each use, assuring the reproducibility of the particle counter by permitting a continuous record of its performance using a NIST™ traceable microsphere suspension with a precise concentration.


CC10 and CC25 are ideal for USP 788 and 788 interim testing.


Certifications: USP, Calibration and Traceability to NIST.

Order Now

SPECIFICATIONS

Environmentally Preferable
Appearance White or Colorless
Stability Contains trace amount of surfactant
Shelf life Product stable for a minimum of six months
Storage temperature 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR