Q-SWAB™ Dry Swab with Transport Tube, Simport
Supplier: Simport Scientific
T307-5A T307-10A
76076-724CS 598.17 CAD
76076-724 76076-726
Q-SWAB™ Dry Swab with Transport Tube, Simport
Swabs Swabs for Sampling
Sterile specimen collection swabs.
  • Polypropylene tubes are translucent
  • Cap is equipped with a silicone washer
  • Non-graduated
The Q-Swab™ design is self-standing, but with round bottom. The polyester swab holds securely inside the screw cap, making it ideal for specimen collection and protection from contamination. The tube material make it easy to see all contents. The cap features with washer fitted inside to ensure a positive seal at any temperature.