Scintillation Vials (20 ml), Simport
Supplier: Simport Scientific
S220-1 S220 S220 S220-1
CA73960-366CS 655.59 CAD
CA73960-366 CA73960-368 76299-278 76299-280
Scintillation Vials (20 ml), Simport
Vials
Shoulderless vial features a full width opening of 23 mm for easy access.
  • High degree of resistance to organic solvents
  • Sufficient translucence for viewing contents
  • Easy ¹/₄ turn cap
  • Polyethylene closure may not be autoclaved
The presence of filter paper can be easily detected. The unique thread design of the closure allows the vial to be sealed. The quality of the sealing system is such that it is not necessary to tighten the closure with a lot of pressure to achieve a leakproof seal.