YMC-Pack Pro C8 HPLC Columns, YMC America
Supplier: YMC America,Inc
OS12S03-0502WT OS12S03-0503WT OS12S03-0546WT OS12S03-1002WT OS12S03-1003WT OS12S03-1046WT OS12S03-1502WT OS12S03-1503WT OS12S03-1546WT OS12S03-H502WT OS12S03-H546WT OS12S03-L502WT OS12S03-L546WT OS12S05-0502WT OS12S05-0503WT OS12S05-0546WT OS12S05-1002WT OS12S05-1046WT OS12S05-1501WT OS12S05-1502WT OS12S05-1503WT OS12S05-1506WT OS12S05-1546WT OS12S05-2501WT OS12S05-2503WT OS12S05-2546WT OS12S05-L502WT OS12S05-L546WT OS12S03-0504WT OS12S03-2503WT OS12S05-0504WT OS12S05-1003WT OS12S05-2502WT
97031-880EA 1466.7 CAD
97031-880 97031-884 97031-890 97031-894 97031-898 97031-902 97031-906 97031-910 97031-914 97031-924 97031-926 97031-928 97031-932 97031-950 97031-954 97031-964 97031-968 97031-974 97031-976 97031-980 97031-984 97031-986 97031-990 97031-992 97031-994 97032-000 97032-006 97032-008 97032-016 97032-020 89216-842 89216-844 89216-856 89216-858 89216-860
YMC-Pack Pro C8 HPLC Columns, YMC America
Chromatography Columns
These columns provide excellent peak shape with basic compounds due to the advanced end capping technology employed to reduce residual silanol activity.
  • Consistently reproducible quality
  • Documentation of each manufacturing step
  • Consistently high packing quality and column efficiency
  • Stringent specifications
  • Individual test of each column
  • Lot identification for each column
The ultrapure silica and very low residual silanol activity makes reduces the use of ion-pairing reagents or ion-suppression techniques, making them ideal for LC/MS applications.