EcoSense® ODO200M Optical Dissolved Oxygen and Temperature Instrument with Memory, YSI
Supplier: YSI
601023 601024 601025 601026
76300-000EA 1039.32 CAD
76300-000 76300-002 76300-004 76300-006
EcoSense® ODO200M Optical Dissolved Oxygen and Temperature Instrument with Memory, YSI
Electrochemistry Meters Oxygen Meters
The EcoSense® ODO200M is an optical-based dissolved oxygen meter ideal for DO sampling. Sensors are EPA approved for wastewater compliance reporting of DO.
  • Waterproof micro-USB port for transferring data to PC; no software required
  • Holds 250 data sets; no need to write down data
  • Real-time clock for date/time stamp of saved data
  • User-selectable recalibration prompt provides reminder to calibrate
  • Automatic temperature compensation; manual input for salinity and pressure compensation
  • IP 67 waterproof housing
  • Replaceable optical sensor cap with 12 to 18 month life
  • Auto shutoff function after 30 minutes of inactivity
  • User calibratable and replaceable
  • Sampling
Compared to the EcoSense® ODO200 , the ODO200M features extended memory and simple data transfer with a micro-USB port. Optical benefits include no stirring requirement, no warm-up time, no electrolyte solutions, and no electrode maintenance.
The ODO200M is designed for quick, accurate results in an economical platform. The convenient optical dissolved oxygen sensor and the simple, compact handheld design allow the ODO200M to be used in many applications.
Ordering information: The following ODO200M purchase options are available: ODO200M instrument only; or 76300-002 instrument, hard-sided transport case, and probe with one meter field cable; or 76300-004 instrument, hard-sided transport case, and probe with four meter field cable; or 76300-006 instrument, hard-sided transport case, and probe with 10 meter field cable. One-year instrument, waterproof case, intuitive operation, probe/cable and sensor cap warranty.