Double Scale Vernier Caliper, Walter Stern
Supplier: Walter Stern
602B 602A
25729-666EA 56.01 CAD
25729-666 25729-668
Double Scale Vernier Caliper, Walter Stern
Calipers
Caliper reads on a metric scale from 0 to 120 mm in 1 mm divisions. It also reads on an imperial scale from 0 to 5" in 1/16" divisions.
  • Equipped with a roller adjustment
The caliper is also readable on the vernier scale to 1/128" or to 0.1 mm.