Tuffshield™ HPPE Fiber Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
PF13-XS PF13-S PF13-M PF13-L PF13-XL
CA46100-400CS 390.68 CAD
CA46100-400 CA46100-402 CA46100-404 CA46100-406 CA46100-408
Tuffshield™ HPPE Fiber Gloves, Honeywell Safety
Gloves
TuffShield™ gloves are cut resistant, combined with low linting properties. This makes the gloves excellent for food processing, and other industries where hand protection is essential.
  • PowerCuff™ system prevents cuff roll down or blow-out from repeated laundering
  • Elastic cuff features enhanced chemical resistance with color coding for easy sorting
Superior cut protection, utilizes a proprietary engineered fiber process. Made with 'Spectra like' HPPE and abrasion resistant filament around a fiberglass core, the fiber is lightweight, low linting, and does not easily absorb liquids.
Ergonomically designed for better fit and greater productivity. Even in the most demanding environments, TuffShield™ gloves can be worn hour after hour, featuring lightweight, flexible and breathable material.
Certifications: EN 388: 254X.