NorthFlex Red™ 15 Gauge Nylon Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
NF11X/6XS NF11X/7S NF11X/8M NF11X/9L NF11X/10XL NF11X/11XXL
CA89041-792CS 1025.24 CAD
CA89041-792 CA89041-794 CA89041-796 CA89041-798 CA89041-800 CA89041-802
NorthFlex Red™ 15 Gauge Nylon Gloves, Honeywell Safety
Gloves
Lightweight, glove with foam PVC coating on palm and fingertips.
  • No coating on back of hand to increase ventilation
  • Black PVC foam over red nylon
  • Seamless knit liner