CSM Glovebox Gloves, Honeywell Safety
Supplier: Honeywell Safety
10Y3032A/9Q Y154A/10 Y154A/7 Y154A/8 10Y1532A/9Q 10Y1532A/9Q Y154A/9
CA10746-664PR 1274.92 CAD
CA10746-664 89186-130 94000-888 94000-886 89186-134 89186-132 89186-136
CSM Glovebox Gloves, Honeywell Safety
Gloves
CSM glove has outstanding mechanical properties to protect against abrasion, tears, and ozone and oxidizing chemicals.
  • Multi-dipped construction
  • Easily cleaned
  • Unsupported
The white surface is specially designed to easily identify contamination. CSM material provides superior protection, while also having an inherently soft and comfortable feel for greater worker comfort.
Certifications: ANSI/ISEA 105 - ASTM D3389-05 Abrasion Test. ANSI/ISEA 105 - ASTM D412 - Physical properties. ANSI/ISEA 105 - ASTM F1790-97 Cut Test. EN 374 - Permiation and breakthrough time. EN 388 - Mechanical risks from abrasion, cut, tear and puncture. EN 420 glove standards. OSHA Standard -29 CFR 1915.157.