Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
Supplier: MAPA
CRYKIT550558ZQK CRYKIT550559ZQK CRYKIT550550ZQK CRYKIT550551ZQK
75841-230EA 488.74 CAD
75841-230 75841-232 75841-234 75841-236
Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
Gloves
These gloves are designed for applications handling liquid nitrogen and other cryogenic gases, to protect from cold contact and prevent burns from liquid gas leakage.
  • Multilayer fleece lining
  • Ventilation technology
  • Waterproof
Gloves remain ventilated due to the inner polyolefin porelle membrane which allows for air to flow and keeps hands dry.
A special elastic laminated blue fabric provides dexterity and comfort in the glove and cuff areas.
Certifications: CE Cat. III 0498 marked. Cold Hazards EN511: 231. Mechanical Hazards EN388: 2422.