874 Butyl Rubber Chemical Protection Gloves, Showa
Supplier: Showa-Best Glove
874-10 874-11
CA28888-344PK 113.2 CAD
CA28888-344 CA28888-346
874 Butyl Rubber Chemical Protection Gloves, Showa
Gloves
Engineered with the ultimate combination of protection and dexterity, this glove features a superior resistance to highly corrosive acids and is designed for handling ketones and esters.
  • Unlined butyl rubber with smooth grip
  • Protection against ketones, alcohols, nerve agents
  • Superior resistance to highly corrosive acids
Provides the highest permeation resistance to gases and water vapors of any protective material used to make gloves.
Certifications: CE Cat. III - 0299 marked. EN 388:2003-0010. EN 374-2:2003.