772 Chemical Resistant Extended Sleeve Gloves, Showa
Supplier: Showa
772L-09 772XL-10 772M-08
CA89370-889CS 2611.13 CAD
CA89370-889 CA89370-885 CA89370-887
772 Chemical Resistant Extended Sleeve Gloves, Showa
Gloves
A flexible, robust glove offering great dexterity and a good resistance to tearing. Protects the hand from oils, hydrocarbons, grease and abrasion, with effective long lasting grip.
  • Fully coated, extra coating over entire hand
  • Extended sewn sleeve, elasticated border
  • Rough finish over entire surface
  • Impermeable for working in damp or greasy environments
  • Antibacterial and anti-odor treatment
  • Anatomical shape
  • Cut and sewn
  • Eyelet for ventilation and hanging
Designed for easy movement and continuous wear, the extended sleeve provides protection up to the shoulder and stays correctly adjusted. The rough surface offers a superior grip, while maintaining a tactile feel.
Certifications: CE Cat. III marked. EN 388:2016-4111X. EN ISO 374-1:2016/Type B JKL. EN ISO 374-5:2016.