MagFuge® High SPD Centrifuge/3 L Magnetic Stirrer, Heathrow Scientific
Supplier: Heathrow Scientific
120581 120582
76311-652EA 2032.78 CAD
76311-652 76311-654
MagFuge® High SPD Centrifuge/3 L Magnetic Stirrer, Heathrow Scientific
Centrifuges Microcentrifuges
Industry's first high-speed centrifuge and 3 l magnetic stirrer combined into one unit. Enables a substantial cost savings for labs by not having to purchase two pieces of equipment, a stirrer and a centrifuge, because the MagFuge® does it all.
  • Modern low profile design is compact and has a small foot print
  • Easy to use back-lit digital panel
  • Programable digital control
  • Rotor sensing technology
  • Long-life maintence free brush-less motor
  • Five-year warranty
  • Power source: Universal power cord
The MagFuge® is the industry’s first high-speed centrifuge (12,More Product Information