D-(-)-Fructose, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Supplier: MilliporeSigma
Synonyms:
D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
1.05321.9029
1.05321.9525
1.05321.9525
CA1.05321.9029EA
2822.65
CAD
CA1.05321.9029
CA1.05321.9525
EM1.05321.9525
D-(-)-Fructose, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
D(-)-Fructose
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 100…104 °C (dec.) Density: 1.59 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00148910 CAS Number: 57-48-7 EINECS: 200-333-3 Merck Index: 13,04295 |
Specification Test Results
Assay (α 20/D, 100 g/l, water, calc. on anhydrous substance) | 98.5 - 101.2 % |
Assay (α 25/D, 100 g/l,water, calc. on anhydrous substance) | 98.0 - 102.0 % |
Identity | passes test |
Appearance of solution (EP) | passes test |
Appearance of solution (USP) | passes test |
Acidity or alkalinity | passes test |
Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance) | -93.5 - -91.0 ° |
Spec. rotation (α 25/D; 10 %; water; calc. on anhydrous substance) | -90.8 - -87.2 ° |
Chloride (Cl) | ≤ 0.01 % |
Sulfate (SO₄) | ≤ 0.02 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
As (Arsenic) | ≤ 0.0001 % |
Ba (Barium) | passes test |
Calcium and Magnesium (as Ca) | ≤ 0.005 % |
Cu (Copper) | ≤ 0.0025 % |
Pb (Lead) | ≤ 0.00001 % |
Zn (Zinc) | ≤ 0.0025 % |
Glucose (HPLC) | ≤ 0.5 % |
Foreign sugars | passes test |
5-Hydroxymethylfurfural and related substances | passes test |
Residual solvents (ICH Q3C) | excluded by the production process |
Sulfated ash | ≤ 0.1 % |
Water | ≤ 0.5 % |
Corresponds to Ph Eur, BP, USP, FCC |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...