Water USP for irrigation, sterile

Supplier: NURSE ASSIST, INC.

6210 6250 6260
76285-232CS 107.38 CAD
76285-232 76285-238 76285-240 76285-246
Water USP for irrigation, sterile
Water
Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR