Cultubes™, Sterile Culture Tubes, Simport Scientific
Supplier: Simport Scientific
T425-2 T425-3 T425-6 T425-2A T425-6A T425-33 T426-2 T426-3 T426-6 T426-2A T426-6A T426-33 T425-2 T425-2A T425-3 T425-33 T425-6 T426-33 T426-6 T426-6A T425-6A T426-2 T426-2A T426-3
CA10798-856CS 299.5 CAD
CA10798-856 CA10798-858 CA10798-860 CA10798-862 CA10798-864 CA10798-866 CA10798-868 CA10798-870 CA10798-872 CA10798-874 CA10798-876 CA10798-878 76299-300 76299-302 76299-322 76299-324 76299-326 76299-328 76299-330 76299-332 76299-372 76299-374 76299-376 76299-378
Cultubes™, Sterile Culture Tubes, Simport Scientific
Tubes Culture Tubes
Cultubes™ are manufactured with precision molding using virgin thermoplastics to ensure that all tubes are uniform in size and shape.
  • Disposable sterile tubes
  • Biologically inert and exampt from mold release agents
These disposable, non-graduated, sterile culture tubes can be used for most routine laboratory procedures. They are biologically inert and exempt from mold release agents. Precision molding with virgin thermoplastics ensures that the tubes will be uniform in size and shape. High resistance to breakage reduces danger in handling infectious or other potentially harmful cultures.
Transparent polystyrene tubes will withstand moderate centrifugation speeds (1400×g), and temperatures to 70 °C. Translucent polypropylene tubes can be centrifuged at higher speeds (3000×g), and resist temperatures from –190 to +120 °C.
Delivery information: Tubes are supplied with either a two-position ribbed polyethylene cap (which can be left loose for aerobic work or sealed for anaerobic cultures), or they are supplied without caps for general-purpose work.