xPIPETTE™ Adjustable Volume Ergonomic Pipettes, Single and Multichannel, Biotix
Supplier: Biotix
30459891 30459892 30459893 30459894 30459895 30459890 30551495 30537396 30537397 30537449 30537450 30537451 30537452 30537454 30537455
76294-096EA 665.61 CAD
76294-096 76294-098 76294-100 76294-102 76294-104 76294-106 76409-446 76409-448 76409-450 76409-452 76409-454 76409-456 76409-458 76409-460 76409-462
xPIPETTE™ Adjustable Volume Ergonomic Pipettes, Single and Multichannel, Biotix
Pipettes Micropipettes
xPIPETTES™ offer accuracy and precision with low ejection forces. Biotix uses patented tip ejection technology to reduce fatigue and makes the xPIPETTE a top choice to improve your lab’s ergonomics.
  • Fully compatible with Rainin LTS tips and Biotix xTIP4™ tips
  • Calibrated to ISO 8655 standards with Biotix xTIP4 tips
  • Light spring forces produce a smooth and controlled aspiration with perfect balance
  • Padded ejector button to reduce hand vibrations during tip ejection
  • Simple volume adjustment with a secure lock to prevent volume drift
  • Available in single, 8, and 12-channel models with a symmetrical design for ambidextrous use
These pipettes are designed with durable and lightweight components. They deliver a precise and accurate liquid transfer while maintaining optimal hand comfort.
Certifications: ISO 8655 certified. Calibration certificate included in the box with each pipette. Unique serial ID number under the finger hook for GLP/GMP compliance.
Caution: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.