Nitrosafe Chemical Gauntlet, Portwest
Supplier: Portwest
A810GNRXL A810GNRM A810GNRS A810GNRXXL A810GNRL
76103-712EA 5.28 CAD
76103-712 76103-714 76103-716 76103-718 76103-710
Nitrosafe Chemical Gauntlet, Portwest
Gloves
The Nitrosafe Chemical Gauntlet is chemical resistant, making it suitable for oil and chemical industries.
  • Chemical resistant gauntlet
  • Smooth nitrile coating
  • Fully coated for maximum liquid protection
The textured pattern on the palm provides an excellent grip. Cotton flock linining contributes to added comfort for the wearer.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .10.1. EN 374 (J.K.L). EN 388:2016 - 3.1.0.1.X. ANSI/ISEA 105 - 2011 5.2.1 Chemical Permeation Resistance. EN 420.