Vis-Tex Cut Resistant Gloves, Portwest
Supplier: Portwest
A625O8RL A625O8RM A625O8RXL
76103-658EA 15.09 CAD
76103-658 76103-660 76103-662
Vis-Tex Cut Resistant Gloves, Portwest
Gloves
Offers both cut and heat protection, while maintaining excellent grip properties.
  • High-visibility liner
  • 100% breathable seamless liner
  • HPPE construction
  • Tested for both cut and heat protection (contact heat resistance)
The PU foam coating provides an excellent grip in both wet and dry conditions. Concentrated on the palms so that dexterity is not limited and gloves remain well ventilated.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. ANSI/ISEA 105 - 2016 CUT Level (A4). EN 388:2003 - 4.5.4.3. EN 388:2016 - 4.X.4.3.D. EN 407 X.1.X.More Product Information