MR Cut PU Palm Gloves, Portwest
Supplier: Portwest
A622G7RL A622G7RM A622G7RXL A622G7RXXL
76103-650EA 12.31 CAD
76103-650 76103-652 76103-654 76103-656
MR Cut PU Palm Gloves, Portwest
Gloves
Level C cut resistant glove for protection against cuts and sharp objects. Seamless liner for added comfort and dexterity.
  • Level C cut resistance
  • 13 gauge liner for a perfect fit
  • Smooth PU coating for increased abrasion resistance
  • Palm dipped to increase dexterity and ventilation
  • Breathable seamless liner
  • Medium risk cut protection
  • Superb abrasion and tear resistance
EN 407 contact heat level 1 provides short term protection against temperatures up to 100 °C.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. EN 388:2016 - 4.X.4.3.C. EN 407 X.1.X.X.X.X. ANSI/ISEA 105 - 2011 Cut Level (A2). EN 420. EN 388:2003 - 4.5.4.3. ANSI cut level A3.