PVC Knit wrist Gloves, Portwest
Supplier: Portwest
A400BKRL A400BKRM A400BKRXL A400BKRXXL
76103-616EA 6.49 CAD
76103-616 76103-618 76103-620 76103-622
PVC Knit wrist Gloves, Portwest
Gloves
This fully dipped glove allows for advanced water and oil protection.
  • Fully coated for maximum liquid protection
  • Phthalate-free
  • Extremely flexible PVC
  • Superb abrasion and tear resistance
  • Knitted cuff for comfort and warmth
PVC knitwrist fully dipped working glove with knitted wrist and cotton interlock lining. It has a smooth finish with the highest abrasion resistance rating.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 ABRASION Level (3). EN 388:2016 - 4.1.2.1.X. EN 420. REACH Compliant.