Hi-Vis Grip Gloves, Portwest
Supplier: Portwest
A340YERL A340YERM A340YERXL
76103-598EA 5.17 CAD
76103-598 76103-600 76103-602
Hi-Vis Grip Gloves, Portwest
Gloves
Offering the highest levels of comfort from foam technology. The latex foam coating provides an excellent grip in both wet and dry conditions.
  • Palm dipped to increase dexterity and ventilation
  • 100% breathable seamless nylon lining
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 ABRASION Level (2). EN 388:2003 - 2.1.2.1. EN 420.