Anti Vibration Gloves, Portwest
Supplier: Portwest
A790BKRL A790BKRM A790BKRXL
76103-704EA 52.23 CAD
76103-704 76103-706 76103-708
Anti Vibration Gloves, Portwest
Gloves
Specially designed to reduce the effects of impact and vibration when using jack hammers, concrete breakers etc.
  • 100% breathable seamless cotton/nylon liner
Anti-vibration pods covering palms are made of specially formulated rubber chloroprene.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. EN388:2016 - 3.1.4.2.X. EN420. EN10819 (TRm=0.62 TRh=0.55) vibration and shock protection. Meets anti-vibration glove standards EN 10819.