Anti Impact Cut Resistant Gloves, Portwest
Supplier: Portwest
A722G8RL A722G8RM A722G8RXL
76103-680EA 38.32 CAD
76103-680 76103-682 76103-684
Anti Impact Cut Resistant Gloves, Portwest
Gloves
Anti Impact Cut Resistant Gloves combines superior cut protection and maximum impact protection using TPR protective pods across the back of the hand.
  • ANSI cut resistance level A4
  • Reinforced thumb crotch
  • Padded palms
  • Hook and loop strap for safe secure fit
  • HPPE, glass fibre and polyester liner material
  • 100% breathable seamless liner
Palms are dipped in a nitrile sandy coating, providing exceptional grip in water, grease or oil contact.
Certifications: AS/NZS 2161 .3. AS/NZS 2161 .2. ANSI/ISEA 105 - 2016 CUT Level (A4). EN 388:2003 - 4.5.4.3. EN 388:2016 - 2.1.4.2.X. CE marked. EN 420.