Flexo Grip Nitrile Gloves, Portwest
Supplier: Portwest
A310GRWL A310GRWM A310GRWXL A310GRWXXL
76103-558EA 2.29 CAD
76103-558 76103-560 76103-562 76103-564
Flexo Grip Nitrile Gloves, Portwest
Gloves
The smooth nitrile coating offers increased abrasion resistance, making these gloves ideal for use auto repair and construction.
  • Palm dipped to increase dexterity and ventilation
  • 100% breathable seamless pylon lining
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 ABRASION Level (2). EN4 20. EN 388:2003 - 4.1.2.2.
Order Now
SPECIFICATIONS
Model A310
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Latex/Latex-free Latex-free
Material Nitrile
Finish Smooth Palm Coating