SensiCare® Powder-Free Stretch Vinyl Sterile Exam Gloves, Medline
Supplier: Medline
484402 484403 484406
CA54720-011CS 477.88 CAD
CA54720-011 CA54720-012 CA54720-015
SensiCare® Powder-Free Stretch Vinyl Sterile Exam Gloves, Medline
Gloves
SensiCare® sterile stretch vinyl exam gloves feature an advanced formulation polyvinyl chloride material for better tactile sensitivity and fit.
  • Beaded cuff
  • No powder or chemical accelerators
  • Available in singles and pairs
The plastic packaging is easy to open, moisture resistant and minimizes fiber tears. It's smaller size requires minimal storage space.
Glove color: Beige
Order Now
SPECIFICATIONS
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Finish Smooth
Cuff style Beaded
Finger thickness 5.9 mil
Palm thickness 5.1 mil
Sterility Sterile