Cryo-Grip® Waterproof Gloves, Tempshield®
Supplier: Tempshield
CGSHLWP-CS CGSHXLWP-CS CGWRSWP-CS CGWRMWP-CS CGWRLWP-CS CGWRXLWP-CS CGMASWP-CS CGMAMWP-CS CGMALWP-CS CGMAXLWP-CS CGEBSWP-CS CGEBMWP-CS CGEBLWP-CS CGEBXLWP-CS CGSHSWP-CS CGSHMWP-CS
76180-968CS 6615.16 CAD
76180-968 76180-970 76180-972 76180-974 76180-976 76181-138 76181-140 76181-142 76181-144 76181-146 76181-148 76181-150 76181-152 76181-154 76181-156 76181-158
Cryo-Grip® Waterproof Gloves, Tempshield®
Gloves
Waterproof Cryo-Grip® gloves offer improved grip on small objects while protecting hands and arms when working in hazardous, ultra-cold environments.
  • Maximum thermal protection
  • Lightweight and comfortable
  • Excellent dexterity
  • Breathable and stay-dry lining
Gloves are designed to provide a high level of thermal protection where the exposure to cryogenic fluids might exist. Made from state-of-the-art materials, these waterproof cryogenic gloves are made to wick away moisture and keep wearer hands dry, to work in comfort for extended periods of time.
Where other gripping materials crack and break in ultra-low environments, the innovative Cryo-Grip® fabric will stay flexible, even in extreme conditions. The multi-layered construction allows for a maximum level of grip, dexterity, flexibility, and thermal protection - essential features when function is important and safety is critical.
Certifications: CE marked. Cat III. EN 420. EN 511. EN 388.