Aurelia® Quest 2.2® Flexible Nitrile Powder-Free Examination Gloves, Supermax
Supplier: Supermax Healthcare Canada
Aurelia
92935
CA76093-785CS 290.34 CAD
CA76093-785
Aurelia® Quest 2.2® Flexible Nitrile Powder-Free Examination Gloves, Supermax
Gloves
Aurelia® Quest 2.2® gives the wearer even greater tactile sensitivity, comfort and flexibility while keeping the fit and look of latex. Ultra-sheer yet durable.
  • 100% latex-free
  • Food contact approved
  • Finger-textured
  • Beaded cuff for ease of donning
Certifications: CE marked. AQL 1.5. EN 455 CE Class 1 Medical Grade. EN 420, EN 388 & EN 374. EN 1186 Food Contact Approved.