KeepKleen® Powder-Free Latex Disposable Gloves, Superior Glove®
Supplier: Superior Glove WorksLtd.
RDLPF/L RDLPF/S RDLPF/M RDLPF/XL
CA89077-296CS 267.27 CAD
CA89077-296 CA89077-298 CA89077-300 CA89077-302
KeepKleen® Powder-Free Latex Disposable Gloves, Superior Glove®
Gloves
KeepKleen® gloves are ergonomically shaped with excellent touch sensitivity, making them perfect for dental and medical examinations, food processing, and laboratory work.
  • Food safe
  • No natural rubber proteins that may cause allergies
  • Beaded cuffs help prevent roll back
Ergonomic shape helps to reduce hand fatigue during longer wear periods. The textured grip improves handling while remaining extremely touch sensitive.
Glove color: Clear
Certifications: CFIA approved for food handling. Class 2 medical grade.
Caution: Latex gloves may cause allergic reactions in some individuals.