KeepKleen® Heavy Duty Food Safe Disposable Nitrile Glove, Superior Glove®
Supplier: Superior Glove WorksLtd.
RD8NPF/S RD8NPF/M RD8NPF/L RD8NPF/XL RD8NPF/XXL
CA89087-563BX 58.73 CAD
CA89087-563 CA89087-565 CA89087-567 CA89087-569 CA89087-571
KeepKleen® Heavy Duty Food Safe Disposable Nitrile Glove, Superior Glove®
Gloves
Designed with silicone-free materials to prevent contamination and increase food safety.
  • Beaded cuff for easier donning
  • Provides greater strength and puncture protection
  • Contains no natural rubber proteins that may cause allergies
Certifications: CFIA compliant.
Caution: Not for medical use.