Sterile Latex Surgical Gloves, Safecross First Aid
Supplier: Safecross First Aid Limited
18371 18372 18373
CA18371EA 198.09 CAD
CA18371 CA18372 CA18373
Sterile Latex Surgical Gloves, Safecross First Aid
Gloves
Surgical-grade, disposable latex medical gloves prevent direct contact with blood and other bodily fluids.
  • Embossed texture allows for better grip and increased comfort
Glove color: Clear
Certifications: Meets or exceeds CGSB requirements with a minimum AQL of 1.5 for freedom of holes.
Packaging: Individually wrapped in pairs (left and right hands).
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Disposable
Latex/Latex-free Latex
Powdered/Powder-free Powdered
Finish Embossed
Finger thickness 3.9 mil
Sterility Sterile